2,2-Dipropylpentanenitrile
Synonym(s):
2,2-Dipropylvaleronitrile, 4-Cyano-4-propylheptane, NSC 915, Tripropylacetonitrile
Empirical Formula (Hill Notation):
C11H21N
CAS Number:
Molecular Weight:
167.29
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing