![2,2-Difluoro-1,3-benzodioxole-4-carbaldehyde AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/197/827/8c651b63-61e5-4b2e-a927-4b366d720af5/640/8c651b63-61e5-4b2e-a927-4b366d720af5.png)
2,2-Difluoro-1,3-benzodioxole-4-carbaldehyde
![2,2-Difluoro-1,3-benzodioxole-4-carbaldehyde AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/197/827/8c651b63-61e5-4b2e-a927-4b366d720af5/640/8c651b63-61e5-4b2e-a927-4b366d720af5.png)
Synonym(s):
2,2-Difluoro-1,3-benzodioxole-4-carboxaldehyde, 2,2-Difluoro-1,3-benzodioxole-4carbaldehyde, 2,2-Difluorobenzo[d][1,3]dioxole, 4-Formyl-2,2-difluoro-1,3-benzodioxole
Empirical Formula (Hill Notation):
C7H4F2O2
CAS Number:
Molecular Weight:
158.10
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing