Skip to Content
Merck
2,2′-Bipyridine-4,4′-dicarboxylic acid 98%

2,2′-Bipyridine-4,4′-dicarboxylic acid

2,2′-Bipyridine-4,4′-dicarboxylic acid 98%
Synonym(s):
2,2′-Bipyridyl-4,4′-dicarboxylic acid
Empirical Formula (Hill Notation):
C12H8N2O4
CAS Number:
Molecular Weight:
244.20

Products

Brand
Formula Weight
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing