![2,2′,3,4,4′,5,5′-Heptachlorobiphenyl (IUPAC No. 180) BCR®, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/412/425/358cc7c9-5a53-4805-aaa3-5d85737871ea/640/358cc7c9-5a53-4805-aaa3-5d85737871ea.png)
2,2′,3,4,4′,5,5′-Heptachlorobiphenyl (IUPAC No. 180)
![2,2′,3,4,4′,5,5′-Heptachlorobiphenyl (IUPAC No. 180) BCR®, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/412/425/358cc7c9-5a53-4805-aaa3-5d85737871ea/640/358cc7c9-5a53-4805-aaa3-5d85737871ea.png)
Synonym(s):
2,2′,3,4,4′,5,5′-Heptachlorobiphenyl
Empirical Formula (Hill Notation):
C12H3Cl7
CAS Number:
Molecular Weight:
395.32
Products
Formula Weight
Manufacturer
Agency
Format
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing