![2-(1H-1,2,4-Triazole-1-yl)acetic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/620/866/c4309eed-0562-4b51-a659-f9f5077c5590/640/c4309eed-0562-4b51-a659-f9f5077c5590.png)
2-(1H-1,2,4-Triazole-1-yl)acetic acid
![2-(1H-1,2,4-Triazole-1-yl)acetic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/620/866/c4309eed-0562-4b51-a659-f9f5077c5590/640/c4309eed-0562-4b51-a659-f9f5077c5590.png)
Synonym(s):
1,2,4-Triazol-1-ylacetic acid, 1,2,4-Triazole-1-acetic acid
Empirical Formula (Hill Notation):
C4H5N3O2
CAS Number:
Molecular Weight:
127.10
Products
Brand
Formula Weight
Manufacturer
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing