Skip to Content
Merck
(1S)-(−)-Camphanic chloride 98%

(1S)-(−)-Camphanic chloride

(1S)-(−)-Camphanic chloride 98%
Synonym(s):
(−)-Camphanoyl chloride, (1S)-3-Oxo-4,7,7-trimethyl-2-oxabicyclo[2.2.1]heptane-1-carbonyl chloride
Linear Formula:
C10H13O3Cl
CAS Number:
Molecular Weight:
216.66
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing