1-Oleoyl-sn-glycero-3-phosphocholine
Synonym(s):
(7R,18Z)-4,7-Dihydroxy-N,N,N-trimethyl-10-oxo-3,5,9-trioxa-4-phosphaheptacos-18-en-1-aminium 4-oxide, inner salt, 1-(9Z-Octadecenoyl)-sn-glycero-3-phosphocholine, 1-cis-9-Octadecenoyl-sn-glycero-3-phosphocholine, 1-Oleoyl-2-hydroxy-sn-glycerol-3-phosphocholine, 1-Oleoyl-sn-glycero-3-phosphorylcholine, 3-sn-Lysophosphatidylcholine, 1-oleoyl, L-γ-Oleoyl-α-lysolecithin, Lysolecithin, oleoyl, PC(18:1(9Z)/0:0), PC(18:1/0:0)
Empirical Formula (Hill Notation):
C26H52NO7P
CAS Number:
Molecular Weight:
521.67
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Functional Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing