![1-Hexadecanol 95%](/deepweb/assets/sigmaaldrich/product/structures/140/974/4580dc3b-2b03-42c7-97b5-0b43cde49b47/640/4580dc3b-2b03-42c7-97b5-0b43cde49b47.png)
1-Hexadecanol
![1-Hexadecanol 95%](/deepweb/assets/sigmaaldrich/product/structures/140/974/4580dc3b-2b03-42c7-97b5-0b43cde49b47/640/4580dc3b-2b03-42c7-97b5-0b43cde49b47.png)
Synonym(s):
Cetyl alcohol, Palmityl alcohol
Linear Formula:
CH3(CH2)15OH
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Showing 1-3 of 3
Compare
Product Number
Description
Pricing