![1-Fluoro-2,4,6-trimethylpyridinium triflate 95%](/deepweb/assets/sigmaaldrich/product/structures/333/512/7515f3a9-19ee-4db7-a2a4-3a94ca5872bd/640/7515f3a9-19ee-4db7-a2a4-3a94ca5872bd.png)
1-Fluoro-2,4,6-trimethylpyridinium triflate
![1-Fluoro-2,4,6-trimethylpyridinium triflate 95%](/deepweb/assets/sigmaaldrich/product/structures/333/512/7515f3a9-19ee-4db7-a2a4-3a94ca5872bd/640/7515f3a9-19ee-4db7-a2a4-3a94ca5872bd.png)
Synonym(s):
1-Fluoro-sym-collidinium triflate
Empirical Formula (Hill Notation):
C9H11F4NO3S
CAS Number:
Molecular Weight:
289.25
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing