Skip to Content
Merck
1-Ethyl-3-methylimidazolium diethyl phosphate ≥98.0%

1-Ethyl-3-methylimidazolium diethyl phosphate

1-Ethyl-3-methylimidazolium diethyl phosphate ≥98.0%
Synonym(s):
EMIM DEP
Empirical Formula (Hill Notation):
C10H21N2O4P
CAS Number:
Molecular Weight:
264.26

Products

Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-2 of 2