Skip to Content
Merck
1-Decene analytical standard

1-Decene

1-Decene analytical standard
Linear Formula:
CH3(CH2)7CH=CH2
CAS Number:
Molecular Weight:
140.27
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique