Skip to Content
Merck
1-Aza-15-crown-5 97%

1-Aza-15-crown-5

1-Aza-15-crown-5 97%
Synonym(s):
1,4,7,10-Tetraoxa-13-azacyclopentadecane
Empirical Formula (Hill Notation):
C10H21NO4
CAS Number:
Molecular Weight:
219.28
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing