1-Adamantanethiol
Synonym(s):
1-AD, 1-AdSH, 1-AdmSH
Empirical Formula (Hill Notation):
C10H16S
CAS Number:
Molecular Weight:
168.30
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing