Skip to Content
Merck
1,8-Diazabicyclo[5.4.0]undec-7-ene puriss., ≥99.0% (GC)

1,8-Diazabicyclo[5.4.0]undec-7-ene

1,8-Diazabicyclo[5.4.0]undec-7-ene puriss., ≥99.0% (GC)
Synonym(s):
2,3,4,6,7,8,9,10-Octahydropyrimidol[1,2-a]azepine, DBU, 1,8-Diazabicyclo[5.4.0]undec-7-ene, 2,3,4,6,7,8,9,10-Octahydropyrimidol[1,2-a]azepine
Empirical Formula (Hill Notation):
C9H16N2
Molecular Weight:
152.24
EC Number:

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade