Skip to Content
Merck
1,4-Butanedithiol ≥97%, FG

1,4-Butanedithiol

1,4-Butanedithiol ≥97%, FG
Synonym(s):
1,4-Dimercaptobutane, Tetramethylene dimercaptan
Linear Formula:
HS(CH2)4SH
CAS Number:
Molecular Weight:
122.25
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing