Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1,2-Distearoyl-
sn
-glycero-3-phosphoethanolamine
Synonym(s):
1,2-Dioctadecanoyl-
sn
-glycero-3-phosphoethanolamine, 3-
sn
-Phosphatidylethanolamine, 1,2-distearoyl, L-β,γ-Distearoyl-α-cephalin
Empirical Formula (Hill Notation):
C
41
H
82
NO
8
P
CAS Number:
1069-79-0
Molecular Weight:
748.07
EC Number:
213-963-9
Products
Product Category
bioactive small molecules (1)
complex lipids (1)
lipids (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Physical Form
Purity
Optical Isomer
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
P3531
1,2-Distearoyl-
sn
-glycero-3-phosphoethanolamine,
≥99%
Expand
View Pricing