Skip to Content
Merck
1-(2-Bromoethyl)naphthalene 97%

1-(2-Bromoethyl)naphthalene

1-(2-Bromoethyl)naphthalene 97%
Empirical Formula (Hill Notation):
C12H11Br
CAS Number:
Molecular Weight:
235.12

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing