Skip to Content
Merck
vibrant-m

11-4

vibrant-m
Synonym(s):
11-4

Products

Product Category
Biological Source
Tissue
Relevant Disease
Taxon
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing