Product Name
Atto 725 BioReagent, suitable for fluorescence, ≥98.0% (HPCE)
Product Number
47156
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
PURITY (HPCE AREA %)≥ 98.0 %
SOLUBILITY (COLOR)Faint Green to Green
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)1 MG/ML IN DMF
EMISSION MAXIMUM752 ± 5 nm
REMARKS ON LUMINESCENCEEXC. 725 NM IN 0.1 M PHOSPHATE PH 7.0