Product Name
2,5-Dichlorothiophene-3-sulfonamide 97%
Product Number
542768
Product Brand
aldrich
CAS Number
53595-68-9
Molecular Weight
232.11
Test
Specification
APPEARANCEWHITE TO TAN POWDER, CRYSTALS, CRYSTALLINE
 POWDER AND/OR CHUNKS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
TITRATION96.5-103.5% (WITH AGNO3 AFTER O2 COMBUSTION)
HIGH PRESSURE LIQUID96.5% (MINIMUM)
 APPROVED JANUARY 10, 2008 RJM