V900885
Peptone from casein
Vetec™, reagent grade, suitable for microbiology
Synonym(s):
Tryptone from casein
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
bovine milk
Quality Level
grade
reagent grade
product line
Vetec™
shelf life
Limited shelf life, expiry date on the label
storage condition
(Tightly closed)
impurities
≥11% total nitrogen (N)
pH
6.5-7.5 (25 °C, 2% in water)
application(s)
microbiology
Looking for similar products? Visit Product Comparison Guide
General description
Peptone from casein is widely used in industrial microbiology as a nutrient source for the growth of microorganisms in large-scale fermentation processes. Peptone from casein, also known as tryptone or peptic casein peptone, is a water-soluble protein hydrolysate obtained from the digestion of casein and used as a nutrient source in microbial culture media. It provides a wide range of nutrients, including amino acids, vitamins, minerals, and peptides, which support the growth of diverse microorganisms. It is particularly useful for the cultivation of fastidious and heterotrophic microbes that require complex nutrient sources.
Application
Peptone from casein is commonly used as an ingredient in many microbial culture media for the growth and identification of bacteria, yeasts, and molds from various pharmaceutical, environmental, and food and beverage samples.
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Xiaohua Dong et al.
Cell reports, 30(8), 2614-2626 (2020-02-27)
Synapses are fundamental to the normal function of the nervous system. Glia play a pivotal role in regulating synaptic formation. However, how presynaptic neurons assemble synaptic structure in response to the glial signals remains largely unexplored. To address this question
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service