Skip to Content
Merck
All Photos(2)

Key Documents

V800375

Sigma-Aldrich

Sodium dichromate dihydrate

LR, ≥99%

Synonym(s):

Sodium bichromate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2Cr2O7 · 2H2O
CAS Number:
Molecular Weight:
298.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

LR

product line

Vetec

Assay

≥99%

form

crystalline powder

pH

3.5 (20 °C, 100 g/L)

mp

91 °C (lit.)

SMILES string

O.O.[Na+].[Na+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

InChI

1S/2Cr.2Na.2H2O.7O/h;;;;2*1H2;;;;;;;/q;;2*+1;;;;;;;;2*-1

InChI key

JYDRNIYTFCBIFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 1B - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 Inhalation

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabelle Hininger et al.
Free radical biology & medicine, 42(12), 1759-1765 (2007-05-22)
Several studies have demonstrated beneficial effects of supplemental trivalent Cr in subjects with reduced insulin sensitivity with no documented signs of toxicity. However, recent studies have questioned the safety of supplemental trivalent Cr complexes. The objective of this study was
Chad M Thompson et al.
Toxicological sciences : an official journal of the Society of Toxicology, 123(1), 58-70 (2011-06-30)
Chronic ingestion of high concentrations of hexavalent chromium [Cr(VI)] in drinking water induces intestinal tumors in mice. To investigate the mode of action (MOA) underlying these tumors, a 90-day drinking water study was conducted using similar exposure conditions as in
Chad M Thompson et al.
Regulatory toxicology and pharmacology : RTP, 64(1), 68-76 (2012-06-19)
In vitro studies on hexavalent chromium [Cr(VI)] indicate that reduced forms of this metal can interact with DNA and cause mutations. Recently, Cr(VI) was shown to induce intestinal tumors in mice; however, Cr(VI) elicited redox changes, cytotoxicity and hyperplasia -
Silvio De Flora et al.
Mutation research, 610(1-2), 38-47 (2006-07-29)
Chromium(VI) compounds are genotoxic in a variety of cellular systems. Their potential carcinogenicity is affected by toxicokinetic patterns restricting bioavailability to certain targets, and by metabolic pathways affecting interaction of chromate-derived reactive species with DNA. Epidemiological data indicate that chromium(VI)
Amanda J Lee et al.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals, 9(2), 103-115 (2004-09-17)
Hexavalent chromium (Cr[VI]) is a genotoxic carcinogen that has been associated with an increased risk of nasal and respiratory tract cancers following occupational exposure. Although the precise mechanism(s) remain to be elucidated, there is evidence for a role of oxidative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service