1266868
USP
Ethyl palmitate
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Ethyl hexadecanoate, Palmitic acid ethyl ester
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
ethyl palmitate
manufacturer/tradename
USP
refractive index
n20/D 1.440 (lit.)
bp
192-193 °C/10 mmHg (lit.)
mp
24-26 °C (lit.)
density
0.857 g/mL at 25 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
CCCCCCCCCCCCCCCC(=O)OCC
InChI
1S/C18H36O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-4-2/h3-17H2,1-2H3
InChI key
XIRNKXNNONJFQO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service