Skip to Content
Merck
All Photos(1)

Key Documents

TA9H97BAF17F

Targetmol Chemicals Inc.

N-Boc-Phe-Leu-Phe-Leu-Phe

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
785.98

Iupac Name

(tert-butoxycarbonyl)-D-phenylalanyl-L-leucyl-D-phenylalanyl-L-leucyl-L-phenylalanine

Inchi Code

1S/C44H59N5O8/c1-28(2)23-33(46-41(53)36(26-31-19-13-9-14-20-31)49-43(56)57-44(5,6)7)38(50)47-35(25-30-17-11-8-12-18-30)40(52)45-34(24-29(3)4)39(51)48-37(42(54)55)27-32-21-15-10-16-22-32/h8-22,28-29,33-37H,23-27H2,1-7H3,(H,45,52)(H,46,53)(H,47,50)(H,48,51)(H,49,56)(H,54,55)/t33-,34-,35+,36+,37-/m0/s1

InChI key

NGNZQSPFQJCBJQ-NSDPJUTJSA-N

storage temp.

-80/-20

Purity

100%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/N-Boc-Phe-Leu-Phe-Leu-Phe

MSDS

https://www.targetmol.com/compound/N-Boc-Phe-Leu-Phe-Leu-Phe


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service