Skip to Content
Merck
All Photos(1)

Key Documents

SY3H6E4112FB

Synthonix Corporation

(6-Chloro-4-methoxypyridin-3-yl)methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
173.6
MDL number:

Iupac Name

(6-chloro-4-methoxy-3-pyridinyl)methanol

Inchi Code

1S/C7H8ClNO2/c1-11-6-2-7(8)9-3-5(6)4-10/h2-3,10H,4H2,1H3

InChI key

STELNDMAIYYQSV-UHFFFAOYSA-N

storage temp.

Room Temperature

Purity

95%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Solid

Product Link

https://synthonix.com/63126.html

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

MSDS

https://synthonix.com/63126.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service