Skip to Content
Merck
All Photos(1)

Key Documents

813076

TSKgel® SP-NPR (2.5 µm) HPLC Columns

L x I.D. 3.5 cm × 4.6 mm, sulfonic acid phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
Pricing and availability is not currently available.

Product Name

TSKgel® SP-NPR HPLC Column, phase sulfonic acid, L × I.D. 3.5 cm × 4.6 mm, 2.5 μm particle size

material

PEEK

product line

TSKgel®

feature

counter ion Na+

manufacturer/tradename

Tosoh SP-NPR, 13076

parameter

≤1 M salt concentration
≤20% organic solvent
1.6 mL/min flow rate
10-60 °C temperature
200 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

3.5 cm × 4.6 mm

matrix

polymer particle platform

matrix active group

sulfonic acid phase

particle size

2.5 μm

operating pH

2-12

separation technique

cation exchange

General description

TSKgel SP-NPR columns are packed with non-porous hydrophilic polymer beads which are surface modified with a strong cation exchanger They are used for the separation and analysis of proteins and peptides. They are particularly useful for adeno-associated viruses and other large macromolecules.

Physical form

Shipped in DI water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Protocols

HPLC Analysis of Purified Adeno-associated Virus (AAV) on TSKgel® SP-NPR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service