Skip to Content
Merck
All Photos(1)

Documents

48611

Supelco

Trichlorofluoromethane solution

200 μg/mL in methanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

FC(Cl)(Cl)Cl

InChI

1S/CCl3F/c2-1(3,4)5

InChI key

CYRMSUTZVYGINF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoping Zhang et al.
Journal of hazardous materials, 186(2-3), 1816-1822 (2011-01-11)
Adsorption on carbon fixed-beds is considered as an inexpensive and highly effective way for controlling chlorofluorocarbons (CFCs) emissions. In the present work, a dynamic model under constant-pattern wave conditions has been developed to predict the breakthrough behavior of trichlorofluoromethane (CFC-11)
Yih-jin Tsai et al.
Journal of environmental sciences (China), 17(5), 838-841 (2005-11-30)
Compounds in the atmosphere contaminate samples of groundwater. An inexpensive and simple method for collecting groundwater samples is developed to prevent contamination when the background concentration of contaminants is high. This new design of groundwater sampling device involves a glass
U E Krone et al.
FEMS microbiology letters, 69(2), 201-204 (1992-01-01)
Methanobacterium barkeri was found to catalyze the reductive dehalogenation of trichlorofluoromethane (CFC-11), also known as FREON 11. Products detected were CHFCl2, CH2FCl, CO and fluoride.
Abdennour Bouhroum et al.
International journal of pharmaceutics, 391(1-2), 98-106 (2010-02-27)
The aims of this study were to investigate and characterize the physico-chemical properties of beclomethasone dipropionate (BDP) crystallized from tricholoromonofluoromethane (CFC-11). Physical interactions in a model pressurised metered dose inhaler (pMDI) system and changes in surface energy after size reduction
T Kyrklund et al.
Scandinavian journal of work, environment & health, 14(2), 91-94 (1988-04-01)
Organic solvents are often present as mixtures in various industrial and house-hold products. The adverse effects arising from exposure to these solvents have often been generalized to concern the whole group of solvents. In an examination of the possibility that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service