Documents
33136
Supelco
Crimp seals with PTFE/rubber septa
seal diam. 8 mm, open center, silver aluminum seal, PTFE/red rubber septum, pkg of 1000 ea
About This Item
UNSPSC Code:
41122001
material
PTFE/red rubber septum
silver aluminum seal
feature
open center
packaging
pkg of 1000 ea
seal diam.
8 mm
Choose from one of the most recent versions:
Certificates of Analysis (COA)