Skip to Content
Merck
All Photos(1)

Documents

Z744097

Cool Brick

parameter at -10 °C

Synonym(s):

Cooling Box, dry block, tube cooling block, cooling block

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103025

packaging

pack of 1 ea

manufacturer/tradename

Diversified Biotech BRIK-1520

parameter

at -10 °C (stable for 10 hours)
at -10 °C

Looking for similar products? Visit Product Comparison Guide

General description

Each Cool Brick allows up to 10 hours of benchtop temperature stability. For maximum cooling results freeze bricks overnight: 1°C Brick in standard freezer, -10°C Brick in a -80°C Freezer. Multiple Bricks can be used in rotation to maintain desired sample temperature.

Application

PCR

Features and Benefits

Up to 10 hours of benchtop temperature stability

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service