Skip to Content
Merck
All Photos(4)

Key Documents

Z742616

PAGE-Optimizer Sample Preparation Column

30 columns

Sign Into View Organizational & Contract Pricing

Select a Size

30 EA
€125.00

€125.00


Estimated to ship on16 April 2025



Select a Size

Change View
30 EA
€125.00

About This Item

UNSPSC Code:
41123100
NACRES:
NA.56

€125.00


Estimated to ship on16 April 2025


feature

30 columns

packaging

pkg of 30 ea

manufacturer/tradename

G-BioSciences 786-948

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

PAGE-Optimizer is a unique, spin column for the preparation of samples for SDS-PAGE. The columns contain a proprietary separation matrix that removes contaminants such as salts, detergents, cellular agents and other common buffering agents. PAGE-Optimizer helps eliminate smeared lanes, distorted and/ or smiling bands and swollen lanes. PAGE-Optimizer has also been shown to reveal protein bands previously masked by the above interfering agents.

Includes: 30 columns

Legal Information

PAGE-Optimizer is a trademark of Geno Technology, Inc.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service