Skip to Content
Merck
All Photos(1)

Documents

T9514

Sigma-Aldrich

Tetrahydropapaverine hydrochloride

≥90%, powder

Synonym(s):

6,7-Dimethoxy-1-(3,4-dimethoxybenzyl)-1,2,3,4-tetrahydroisoquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25NO4 · HCl
CAS Number:
Molecular Weight:
379.88
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

≥90%

form

powder

color

white

solubility

H2O: soluble

storage temp.

2-8°C

SMILES string

Cl.COc1ccc(CC2NCCc3cc(OC)c(OC)cc23)cc1OC

Biochem/physiol Actions

Phosphodiesterase inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Suzuki et al.
Advances in neurology, 53, 215-218 (1990-01-01)
Effects of MPTP-like compounds on mitochondrial respiration, activity of NADH-ubiquinone oxidoreductase (complex I) and on ATP synthesis were reported. Mitochondria prepared from mouse whole brains were used. The compounds tested include tetrahydroisoquinoline (TIQ), tetrahydropapaveroline (THP), tetrahydropapaverine (THPV), and salsolinol. TIQ
S Chulia et al.
British journal of pharmacology, 122(3), 409-416 (1997-11-14)
1. In the present work, the properties of 3-methyl isoquinoline, 3,4-dihydropapaverine, tetrahydropapaverine and tetrahydropapaveroline were compared with those of papaverine and laudanosine. The work includes functional studies on rat isolated aorta contracted with noradrenaline, caffeine or KCl, and a determination
K Suzuki et al.
Neurochemical research, 15(7), 705-710 (1990-07-01)
Since the discovery of 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP)-induced parkinsonism, it has been postulated that (a) MPTP-like toxin(s) such as 1,2,3,4-tetrahydroisoquinoline (TIQ) may induce Parkinson's disease. As the neuronal degeneration in MPTP-induced parkinsonism is thought to be caused by the inhibition of the
F S Messiha
Pharmacology, biochemistry, and behavior, 35(1), 29-33 (1990-01-01)
The in vivo effect of amantadine, chlorpromazine and reserpine on testicular aldehyde dehydrogenase (T-ALDH) was studied as a function of mouse strain. The effect of Leu-enkephalin and tetrahydropapaverine on rodent T-ALDH was also studied in vivo. The in vitro effect
Yunjo Soh et al.
Biological & pharmaceutical bulletin, 26(6), 803-807 (2003-06-17)
Protective effects of quinic acids from Aster scaber on tetrahydropapaveroline (THP)-induced cell toxicity were evaluated in rat C6 glioma cells. Among 4 quinic acid derivatives tested, (-) 4,5-dicaffeoyl quinic acid (QA3) exhibited the highest protective effect against THP-induced cell toxicity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service