Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T4781

Sigma-Aldrich

1,4,7-Triazacyclononane

~97%

Synonym(s):

Octahydro-1H-1,4,7-triazonine, Triethylenetriamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15N3
CAS Number:
Molecular Weight:
129.20
Beilstein:
773877
MDL number:
UNSPSC Code:
12350000

Pricing and availability is not currently available.

Assay

~97%

bp

110-130 °C/7 mmHg (lit.)

mp

42-45 °C (lit.)

storage temp.

2-8°C

SMILES string

C1CNCCNCCN1

InChI

1S/C6H15N3/c1-2-8-5-6-9-4-3-7-1/h7-9H,1-6H2

InChI key

ITWBWJFEJCHKSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Possible reagent for compleximetric titrations with high cation-binding selectivity.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service