Skip to Content
Merck
All Photos(1)

Key Documents

SRP5215

Sigma-Aldrich

SMYD3, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

FLJ21080, ZMYND1, ZNFN3A1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~71 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SMYD3(64754)

General description

SMYD3 (SET and MYND domain containing 3) is a histone methyltransferase that plays a role in transcriptional regulation as a member of an RNA polymerase complex. The introduction of SMYD3 enhanced cell growth and the histone methyltransferase activity of SMYD3 involved in the proliferation of cancer cells. A variable number of tandems repeat polymorphism in an E2F-1 binding element in the 5-prime flanking region of SMYD3 is a risk factor for human cancers.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryuji Hamamoto et al.
Nature cell biology, 6(8), 731-740 (2004-07-06)
Colorectal and hepatocellular carcinomas are some of the leading causes of cancer deaths worldwide, but the mechanisms that underly these malignancies are not fully understood. Here we report the identification of SMYD3, a gene that is over-expressed in the majority
Masataka Tsuge et al.
Nature genetics, 37(10), 1104-1107 (2005-09-13)
Histone modification is a crucial step in transcriptional regulation, and deregulation of the modification process is important in human carcinogenesis. We previously reported that upregulation of SMYD3, a histone methyltransferase, promoted cell growth in human colorectal and hepatocellular carcinomas. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service