Skip to Content
Merck
All Photos(1)

Key Documents

SRP0394

Sigma-Aldrich

EZH2/EED/SUZ12/RbAp48 active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥90% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€1,590.00

€1,590.00


Usually ships in 2 weeks.


Select a Size

Change View
50 μG
€1,590.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€1,590.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

48 kDa (RbBP5)
51 kDa (EED)
86 kDa (EZH2)
87 kDa (SUZ12)

packaging

pkg of 50 μg

shipped in

dry ice

storage temp.

−70°C

Gene Information

General description

Complex of human EZH2 (GenBank Accession No. NM_004456), amino acids 2-end with N-terminal His-tag, MW= 86 kDa; human EED (GenBank Accession No. NM_003797) amino acids 2-end with N-terminal Flag-tag, MW= 51 kDa, human SUZ12 (GenBank Accession No. NM_015355) amino acids 2-end with N-terminal His-tag, MW = 87 kDa, and human RbAp48 (GenBank Accession No. NM_005610) amino acids 2-end with N-terminal His-tag, MW = 48 kDa, co-expressed in Baculovirus expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rakotobe, D. et al.
Journal of Virology, 5, 32-32 (2008)
Sooryanarayana Varambally et al.
Science (New York, N.Y.), 322(5908), 1695-1699 (2008-11-15)
Enhancer of zeste homolog 2 (EZH2) is a mammalian histone methyltransferase that contributes to the epigenetic silencing of target genes and regulates the survival and metastasis of cancer cells. EZH2 is overexpressed in aggressive solid tumors by mechanisms that remain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service