Skip to Content
Merck
All Photos(1)

Documents

SML2903

Sigma-Aldrich

KIRA8

≥98% (HPLC)

Synonym(s):

(S)-2-Chloro-N-(6-methyl-5-((3-(2-(piperidin-3-ylamino)pyrimidin-4-yl)pyridin-2-yl)oxy)naphthalen-1-yl)benzenesulfonamide, 2-Chloro-N-[6-methyl-5-[[3-[2-[(3S)-3-piperidinylamino]-4-pyrimidinyl]-2-pyridinyl]oxy]-1-naphthalenyl]benzenesulfonamide, AMG 18, AMG-18, AMG18, Compound 18, IRE1α Kinase-Inhibiting RNase Attenuator 8

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H29ClN6O3S
CAS Number:
Molecular Weight:
601.12
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

KIRA8 is an ATP-competitive, potent and selective IRE1α kinase inhibitor (IRE1α/β Ki = 2/120 nM) that attenuates IRE1α endonuclease activity (IRE1α/β IC50 = 5/55 nM) with good kinome selectivity. KIRA8 inhibits thapsigargin-induced UPR (IC50 = 99 nM; HT1080 XBP1-Luc cells) and shows therapeutic efficacy in murine models of type 1 diabetes (T1D), pulmonary fibrosis, multiple myeloma (MM), and pancreatic neuroendocrine tumors (PanNET) in vivo (30-50 mg/kg/day i.p.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hannah C Feldman et al.
ACS chemical biology, 14(12), 2595-2605 (2019-10-15)
The dual kinase endoribonuclease IRE1 is a master regulator of cell fate decisions in cells experiencing endoplasmic reticulum (ER) stress. In mammalian cells, there are two paralogs of IRE1: IRE1α and IRE1β. While IRE1α has been extensively studied, much less
[Detection of papillomavirus types 6/11, 16/18 and 31/33/35 using DNA/RNA hybridization. Initial experiences].
B Wartusch et al.
Gynakologische Rundschau, 29 Suppl 2, 402-404 (1989-01-01)
Shuhei Morita et al.
Cell metabolism, 25(4), 883-897 (2017-04-06)
In cells experiencing unrelieved endoplasmic reticulum (ER) stress, the ER transmembrane kinase/endoribonuclease (RNase)-IRE1α-endonucleolytically degrades ER-localized mRNAs to promote apoptosis. Here we find that the ABL family of tyrosine kinases rheostatically enhances IRE1α's enzymatic activities, thereby potentiating ER stress-induced apoptosis. During
Maike Thamsen et al.
PloS one, 14(1), e0209824-e0209824 (2019-01-10)
Endoplasmic reticulum stress (ER stress) has been implicated in the pathogenesis of idiopathic pulmonary fibrosis (IPF), a disease of progressive fibrosis and respiratory failure. ER stress activates a signaling pathway called the unfolded protein response (UPR) that either restores homeostasis
Jonathan M Harnoss et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(33), 16420-16429 (2019-08-03)
Multiple myeloma (MM) arises from malignant immunoglobulin (Ig)-secreting plasma cells and remains an incurable, often lethal disease despite therapeutic advances. The unfolded-protein response sensor IRE1α supports protein secretion by deploying a kinase-endoribonuclease module to activate the transcription factor XBP1s. MM

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service