Skip to Content
Merck
All Photos(1)

Key Documents

SML2541

Sigma-Aldrich

JWG-071

≥98% (HPLC)

Synonym(s):

11-(sec-butyl)-2-((2-methoxy-4-(4-(4-methylpiperazin-1-yl)piperidine-1-carbonyl)phenyl)amino)-5-methyl-5,11-dihydro-6H-benzo[e]pyrimido[5,4-b][1,4]diazepin-6-one, 5,11-Dihydro-2-[[2-methoxy-4-[[4-(4-methyl-1-piperazinyl)-1-piperidinyl]carbonyl]phenyl]amino]-5-methyl-11-(1-methylpropyl)-6H-pyrimido[4,5-b][1,4]benzodiazepin-6-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€108.00
25 MG
€360.00

€108.00


Estimated to ship on24 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€108.00
25 MG
€360.00

About This Item

Empirical Formula (Hill Notation):
C34H44N8O3
CAS Number:
Molecular Weight:
612.76
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€108.00


Estimated to ship on24 April 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N6(CCN(CC6)C)C1CCN(CC1)C(=O)c2cc(c(cc2)Nc3nc4[n](c5c([c]([n](c4cn3)C)=O)cccc5)C(CC)C)OC

InChI

1S/C34H44N8O3/c1-6-23(2)42-28-10-8-7-9-26(28)33(44)39(4)29-22-35-34(37-31(29)42)36-27-12-11-24(21-30(27)45-5)32(43)41-15-13-25(14-16-41)40-19-17-38(3)18-20-40/h7-12,21-23,25H,6,13-20H2,1-5H3,(H,35,36,37)

InChI key

ACWOMSOYIIVIRV-UHFFFAOYSA-N

Biochem/physiol Actions

JWG-071 is a selective inhibitor of ERK5 (MAPK7) and LRRK2 with an IC50 value of 88nM for ERK5 inhibition and 109 nM for LRRK2 inhibition.
Selective ERK5 (MAPK7) Inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinhua Wang et al.
ACS chemical biology, 13(9), 2438-2448 (2018-08-14)
Bromodomains have been pursued intensively over the past several years as emerging targets for the development of anticancer and anti-inflammatory agents. It has recently been shown that some kinase inhibitors are able to potently inhibit the bromodomains of BRD4. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service