Skip to Content
Merck
All Photos(1)

Key Documents

SML0357

Sigma-Aldrich

AcH4-21 trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

Ac-SGRGKGGKGLGKGGAKRHRKV trifluoroacetate salt, Ac-Ser-Gly-Arg-Gly-Lys-Gly-Gly-Lys-Gly-Leu-Gly-Lys-Gly-Gly-Ala-Lys-Arg-His-Arg-Lys-Val-OH trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C89H161N37O24 · xC2HF3O2
Molecular Weight:
2133.46 (free base basis)
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95% (HPLC)

form

lyophilized powder

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC(C)C[C@H](NC(=O)[C@H](CCCCN)NC(=O)CNC(=O)CNC(=O)[C@H](CCCCN)NC(=O)[C@H](CCCNC(N)=N)NC(=O)CNC(=O)[C@H](CO)NC(C)=O)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)NCC(=O)NCC(=O)N[C@@H](C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](Cc1c[nH]cn1)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C(C)C)C(O)=O

InChI

1S/C85H155N35O22.C2HF3O2/c1-47(2)36-60(118-76(135)54(22-9-14-30-88)111-66(126)42-102-64(124)40-104-72(131)53(21-8-13-29-87)114-75(134)55(25-17-33-98-83(91)92)112-68(128)44-106-74(133)62(45-121)109-50(6)122)73(132)105-43-67(127)110-52(20-7-12-28-86)71(130)103-39-63(123)101-41-65(125)108-49(5)70(129)113-56(23-10-15-31-89)77(136)116-59(27-19-35-100-85(95)96)79(138)119-61(37-51-38-97-46-107-51)81(140)117-58(26-18-34-99-84(93)94)78(137)115-57(24-11-16-32-90)80(139)120-69(48(3)4)82(141)142;3-2(4,5)1(6)7/h38,46-49,52-62,69,121H,7-37,39-45,86-90H2,1-6H3,(H,97,107)(H,101,123)(H,102,124)(H,103,130)(H,104,131)(H,105,132)(H,106,133)(H,108,125)(H,109,122)(H,110,127)(H,111,126)(H,112,128)(H,113,129)(H,114,134)(H,115,137)(H,116,136)(H,117,140)(H,118,135)(H,119,138)(H,120,139)(H,141,142)(H4,91,92,98)(H4,93,94,99)(H4,95,96,100);(H,6,7)/t49-,52-,53-,54-,55-,56-,57-,58-,59-,60-,61-,62-,69-;/m0./s1

InChI key

PYHKKXHCPQWHEI-ORHMKHTFSA-N

Amino Acid Sequence

Ac-Ser-Gly-Arg-Gly-Lys-Gly-Gly-Lys-Gly-Leu-Gly-Lys-Gly-Gly-Ala-Lys-Arg-His-Arg-Lys-Val-OH trifluoroacetate

Biochem/physiol Actions

AcH4-21 is a good substrate of Protein Arginine Methyltransferases (PRMTs).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service