Skip to Content
Merck
All Photos(1)

Documents

SCP0155

Sigma-Aldrich

GLP-2 1-33 rat / Preproglucagon 126-158 / Glp 2 rat

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C166H256N44O56S1
Molecular Weight:
3796.14
UNSPSC Code:
12352200
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

His-Ala-Asp-Gly-Ser-Phe-Ser-Asp-Glu-Met-Asn-Thr-Ile-Leu-Asp-Asn-Leu-Ala-Thr-Arg-Asp-Phe-Ile-Asn-Trp-Leu-Ile-Gln-Thr-Lys-Ile-Thr-Asp

General description

Glucagon-like peptide 2 (GLP-2), an intestinal specific trophic hormone, is a gastrointestinal peptide with 33% sequence homology to glucagon. L-cells on the mucosa of the small intestine and colon release this enteroendocrine hormone.

Biochem/physiol Actions

Glucagon-like peptide 2 (GLP-2) plays a key role in the stimulation of intestinal epithelial proliferation. It possesses intestinotrophic endocrine/paracrine actions. GLP-2 is essentially a nutrition absorption regulator and an intestinal-specific trophic factor. It exhibits anti-inflammatory effects.
Glycogen-like peptide-2 (GLP-2) is involved in a range of processes in vivo including intestinal growth, reduction in bone breakage appetite and neuroprotection. GLP-2 human and GLP-2 rat may be used to study these and other processes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heather A Redstone et al.
Gastroenterology research and practice, 2010 (2010-10-19)
Background. Glucagon-like peptide 2 (GLP-2) is an intestinal specific trophic hormone, with therapeutic potential; the effects on intestinal healing are unknown. We used a rat model of colonic healing, under normoxic, and stress (hypoxic) conditions to examine the effect of
P L Brubaker et al.
Endocrinology, 138(11), 4837-4843 (1997-11-05)
Glucagon-like peptide-2 (GLP-2) has recently been identified as a stimulator of intestinal epithelial growth, prompting the development of RIA and HPLC methodologies to study this peptide in more detail. A GLP-2-specific antiserum (UTTH-7) was developed that recognizes amino acids 25-30
J Thulesen
Current protein & peptide science, 5(1), 51-65 (2004-02-18)
Glucagon-like peptide 2 (GLP-2) is a newly discovered gastrointestinal peptide with 33% sequence homology to glucagon. GLP-2 has attracted interest because of its potent intestinotrophic endocrine/paracrine actions. The peptide, consisting of 33-amino-acid, results from expression of the glucagon gene in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service