Skip to Content
Merck
All Photos(1)

Key Documents

SAB5300260

Sigma-Aldrich

Monoclonal Anti-LIPA antibody produced in mouse

clone 9G7F12, ascites fluid

Synonym(s):

CESD, LAL, LIPA

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€494.00

€494.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€494.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€494.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

9G7F12, monoclonal

species reactivity

human

technique(s)

direct ELISA: 1:10,000
western blot: 1:500-1:2,000

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LAL(3988)

Immunogen

Purified recombinant fragment of LAL expressed in E.coli.
Mouse monoclonal antibody raised against LAL

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service