SAB5201754
Monoclonal Anti-Tcp1 Alpha - Atto 488 antibody produced in rat
clone 91a, purified immunoglobulin
Synonym(s):
Anti-AI528772, Anti-Ccta, Anti-Tric, Anti-p63
Select a Size
€568.00
Select a Size
About This Item
€568.00
biological source
rat
Quality Level
conjugate
Atto 488 conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
91a, monoclonal
form
buffered aqueous solution
mol wt
antigen predicted mol wt 60 kDa
species reactivity
Drosophila melanogaster, human, mouse, hamster, guinea pig, Saccharomyces cerevisiae, rat, pig, monkey, rabbit, Caenorhabditis elegans, bovine, dog
concentration
1 mg/mL
technique(s)
flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG2a
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
mouse ... Tcp1(21454)
Related Categories
Immunogen
Features and Benefits
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service