Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700671

Sigma-Aldrich

Monoclonal Anti-KRT18-Biotin antibody produced in mouse

clone DC-10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Cytokeratin 18

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€293.00

€293.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€293.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€293.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

DC-10, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... KRT18(3875)

General description

Keratin 18 (KRT 18) or cytokeratin 18 (CK18) encodes the type I intermediate filament chain keratin 18. The antibody DC-10 reacts with Cytokeratin 18 (45 kDa). Cytokeratins are a member of intermediate filaments subfamily represented in epithelial tissues. In human chromosome, the gene KRT18 is localized on 12q13.13.

Immunogen

Human breast carcinoma cell line PMC-42

Application

The reagent is designed for Flow Cytometry. Suggested working dilution is 1:500. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Biochem/physiol Actions

Keratin 18 (KRT18/CK18) pairs with CK8 to form the intermediate filaments and maintain the integrity in hepatocytes. CK18 associates with histone H3 in the progression of hepatocellular carcinoma (HCC). Disorganization or altered levels of CK18 is related to plectin deficiency in HCC. K18 may be a marker for liver diseases. K18 is present in Mallory bodies and is elevated in alcoholic cirrhosis and alcoholic hepatitis. K18 expression is modulated by interleukin 6 (IL-6). K18 interacts with desmosomal protein/ E-cadherin and promotes cell adhesion. CK18 is cleaved by caspases during apoptosis and released into blood as caspase cleaved cytokeratin-18 (CCCK18).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cytokeratin 18-associated Histone 3 Modulation in Hepatocellular Carcinoma: A Mini Review
Lai YCC, et al.
Cancer, Genomics, and Proteomics, 14(4), 219-223 (2017)
TP53 mutation, epithelial-mesenchymal transition, and stem like features in breast cancer subtypes
Coradini D, et al.
BioMed Research International, 2012 (2012)
High serum levels of caspase-cleaved cytokeratin-18 are associated with malignant middle cerebral artery infarction patient mortality
Lorente L, et al.
BMC Neurology, 18(1), 32-32 (2018)
Interleukin-6 Induces keratin expression in intestinal epithelial cells potential role of Keratin-8 in interleukin-6-induced barrier function alterations
Wang L, et al.
The Journal of Biological Chemistry, 282(11), 8219-8227 (2007)
Cell death and prognosis of mortality in alcoholic hepatitis patients using plasma keratin-18
Woolbright BL, et al.
Gene Expression, 17(4), 301-312 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service