Skip to Content
Merck
All Photos(2)

Key Documents

SAB3501074

Sigma-Aldrich

Anti-DNM1L antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€503.00

€503.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€503.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€503.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 81 kDa

species reactivity

mouse, human

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunoblotting: suitable
immunocytochemistry: suitable
immunofluorescence: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... DNM1L(10059)

Immunogen

Antibody was raised against a 19 amino acid peptide near the center of human DNM1L.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3501074.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Frank et al.
Developmental cell, 1(4), 515-525 (2001-11-13)
In healthy cells, fusion and fission events participate in regulating mitochondrial morphology. Disintegration of the mitochondrial reticulum into multiple punctiform organelles during apoptosis led us to examine the role of Drp1, a dynamin-related protein that mediates outer mitochondrial membrane fission.
P Hemachandra Reddy et al.
Brain research reviews, 67(1-2), 103-118 (2010-12-15)
The purpose of this article is to review the recent developments of abnormal mitochondrial dynamics, mitochondrial fragmentation, and neuronal damage in neurodegenerative diseases, including Alzheimer's, Parkinson's, Huntington's, and amyotrophic lateral sclerosis. The GTPase family of proteins, including fission proteins, dynamin
H W Shin et al.
Journal of biochemistry, 122(3), 525-530 (1997-11-05)
The dynamin family of GTP-binding proteins are implicated in vesicular transport. These include mammalian dynamins I, II, III, and yeast Vps1p and Dnm1p. Dynamin is involved in the formation of clathrin-coated vesicles at the plasma membrane. On the other hand
Carola Stockburger et al.
Journal of Alzheimer's disease : JAD, 42(2), 395-411 (2014-06-06)
Recent data suggest that the combined effect of oxidative stress due to aging and slightly elevated amyloid-β (Aβ) levels initiate Alzheimer's disease (AD) long before the clinical onset. Investigations of this early phase are hampered by the lack of cellular
Mabel Lum et al.
International journal of medical microbiology : IJMM, 304(5-6), 530-541 (2014-04-24)
Shigella infection in epithelial cells induces cell death which is accompanied by mitochondrial dysfunction. In this study the role of the mitochondrial fission protein, Drp1 during Shigella infection in HeLa cells was examined. Significant lactate dehydrogenase (LDH) release was detected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service