Skip to Content
Merck
All Photos(1)

Documents

SAB2501166

Sigma-Aldrich

Anti-EBF1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-COE1, Anti-EBF, Anti-FLJ39389, Anti-FLJ41763, early B-cell factor 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EBF1(1879)

Immunogen

Peptide with sequence CHNQLPALANTSV, from the internal region of the protein sequence according to NP_076870.1.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Chromatin immunoprecipitation (1 paper)
Immunoprecipitation (1 paper)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erkang Yi et al.
Clinical and translational medicine, 11(7), e479-e479 (2021-07-30)
Chronic obstructive pulmonary disease is a complex condition with multiple etiologies, including inflammation. We identified a novel long noncoding RNA (lncRNA), interleukin 6 antisense RNA 1 (IL6-AS1), which is upregulated in this disease and is associated with airway inflammation. We
Nikki R Kong et al.
PLoS genetics, 12(2), e1005845-e1005845 (2016-02-24)
Hematopoietic stem cells are capable of self-renewal or differentiation along three main lineages: myeloid, erythroid, and lymphoid. One of the earliest lineage decisions for blood progenitor cells is whether to adopt the lymphoid or myeloid fate. Previous work had shown
Jan-Philipp Mallm et al.
Molecular systems biology, 15(5), e8339-e8339 (2019-05-24)
In chronic lymphocytic leukemia (CLL), a diverse set of genetic mutations is embedded in a deregulated epigenetic landscape that drives cancerogenesis. To elucidate the role of aberrant chromatin features, we mapped DNA methylation, seven histone modifications, nucleosome positions, chromatin accessibility
Greg A Timblin et al.
Journal of immunology (Baltimore, Md. : 1950), 191(9), 4676-4687 (2013-09-27)
The temporal control of RAG (Rag) expression in developing lymphocytes prevents DNA breaks during periods of proliferation that could threaten genomic integrity. In developing B cells, the IL-7R and precursor B cell Ag receptor (pre-BCR) synergize to induce proliferation and
Tobias Strid et al.
Journal of immunology (Baltimore, Md. : 1950), 206(11), 2700-2713 (2021-05-23)
B lymphocyte development is dependent on the interplay between the chromatin landscape and lineage-specific transcription factors. It has been suggested that B lineage commitment is associated with major changes in the nuclear chromatin environment, proposing a critical role for lineage-specific

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service