Skip to Content
Merck
All Photos(2)

Documents

SAB1410958

Sigma-Aldrich

Anti-PFKM antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

GSD7, MGC8699, PFK-1, PFK-M, PFKX

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 85.2 kDa

species reactivity

mouse, human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PFKM(5213)

General description

Phosphofructokinase muscle (PFKM) is a subunit of the mammalian tetramer phosphofructokinase. It is encoded by the gene mapped to human chromosome 12q13.1. PFKM is expressed in muscle, red blood cells, thyroid and brain.
The PFKM gene encodes the muscle isoform of phosphofructokinase (PFK) (ATP:D-fructose-6-phosphate-1-phosphotransferase, EC 2.7.1.11). PFK catalyzes the irreversible conversion of fructose-6-phosphate to fructose-1,6-bisphosphate and is a key regulatory enzyme in glycolysis. Mammalian PFK is a tetramer made up of various combinations of 3 subunits: muscle (PFKM), liver (PFKL; MIM 171860), and platelet (PFKP; MIM 171840), the genes for which are located on chromosomes 12q13, 21q22, and 10p, respectively. The composition of the tetramers differs according to the tissue type. Muscle and liver PFK are a homotetramers of 4M and 4L subunits, respectively. Erythrocytes contain both L and M subunits, which randomly tetramerize to form M4, L4, and M3L, M2L2, and ML3 hybrid forms of the holoenzyme (Vora et al., 1980 [PubMed 6444721]; Raben and Sherman, 1995 [PubMed 7550225]).[supplied by OMIM

Immunogen

PFKM (NP_000280.1, 1 a.a. ~ 780 a.a) full-length human protein.

Sequence
MTHEEHHAAKTLGIGKAIAVLTSGGDAQGMNAAVRAVVRVGIFTGARVFFVHEGYQGLVDGGDHIKEATWESVSMMLQLGGTVIGSARCKDFREREGRLRAAYNLVKRGITNLCVIGGDGSLTGADTFRSEWSDLLSDLQKAGKITDEEATKSSYLNIVGLVGSIDNDFCGTDMTIGTDSALHRIMEIVDAITTTAQSHQRTFVLEVMGRHCGYLALVTSLSCGADWVFIPECPPDDDWEEHLCRRLSETRTRGSRLNIIIVAEGAIDKNGKPITSEDIKNLVVKRLGYDTRVTVLGHVQRGGTPSAFDRILGSRMGVEAVMALLEGTPDTPACVVSLSGNQAVRLPLMECVQVTKDVTKAMDEKKFDEALKLRGRSFMNNWEVYKLLAHVRPPVSKSGSHTVAVMNVGAPAAGMNAAVRSTVRIGLIQGNRVLVVHDGFEGLAKGQIEEAGWSYVGGWTGQGGSKLGTKRTLPKKSFEQISANITKFNIQGLVIIGGFEAYTGGLELMEGRKQFDELCIPFVVIPATVSNNVPGSDFSVGADTALNTICTTCDRIKQSAAGTKRRVFIIETMGGYCGYLATMAGLAAGADAAYIFEEPFTIRDLQANVEHLVQKMKTTVKRGLVLRNEKCNENYTTDFIFNLYSEEGKGIFDSRKNVLGHMQQGGSPTPFDRNFATKMGAKAMNWMSGKIKESYRNGRIFANTPDSGCVLGMRKRALVFQPVAELKDQTDFEHRIPKEQWWLKLRPILKILAKYEIDLDTSDHAHLEHITRKRSGEAAV

Biochem/physiol Actions

Phosphofructokinase muscle (PFKM) functions as a rate-limiting enzyme in glycolysis. Aberrations in the gene expression cause Tarui disease and might also lead to non-insulin-dependent diabetes mellitus (NIDDM). Mutation of the gene might also increases the risk of susceptibility to breast cancer.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Habibul Ahsan et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 23(4), 658-669 (2014-02-05)
Early-onset breast cancer (EOBC) causes substantial loss of life and productivity, creating a major burden among women worldwide. We analyzed 1,265,548 Hapmap3 single-nucleotide polymorphisms (SNP) among a discovery set of 3,523 EOBC incident cases and 2,702 population control women ages
A Kahn et al.
Human genetics, 48(1), 93-108 (1979-04-17)
Isozymic heterogeneity of human phosphofructokinase was investigated by means of ATP inhibition, immunoneutralization by antihuman muscle-type and antiliver-type phosphofructokinase antisera, solubility in (NH4)2SO4 solutions, and starch gel and polyacrylamide slab gel electrophoresis. The enzymes studied by these methods were purified
M Ristow et al.
The Journal of clinical investigation, 100(11), 2833-2841 (1998-02-12)
Non-insulin-dependent diabetes mellitus (NIDDM) is caused by peripheral insulin resistance and impaired beta cell function. Phosphofructo-1-kinase (PFK1) is a rate-limiting enzyme in glycolysis, and its muscle subtype (PFK1-M) deficiency leads to the autosomal recessively inherited glycogenosis type VII Tarui's disease.
T D Howard et al.
Genomics, 34(1), 122-127 (1996-05-15)
Phosphofructokinase (PFK) is a key rate-limiting enzyme in glycolysis and represents a major control point in the metabolism of glucose. There are at least three known isoforms of PFK in humans, referred to as the muscle, platelet, and liver forms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service