Skip to Content
Merck
All Photos(3)

Key Documents

SAB1401130

Sigma-Aldrich

Anti-FABP5 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

E-FABP, EFABP, PA-FABP, PAFABP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€492.00

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€492.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FABP5(2171)

General description

This gene encodes the fatty acid binding protein found in epidermal cells, and was first identified as being upregulated in psoriasis tissue. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism. The human genome contains many pseudogenes similar to this locus. (provided by RefSeq)

Immunogen

FABP5 (NP_001435.1, 1 a.a. ~ 135 a.a) full-length human protein.

Sequence
MATVQQLEGRWRLVDSKGFDEYMKELGVGIALRKMGAMAKPDCIITCDGKNLTIKTESTLKTTQFSCTLGEKFEETTADGRKTQTVCNFTDGALVQHQEWDGKESTITRKLKDGKLVVECVMNNVTCTRIYEKVE

Biochem/physiol Actions

FABPs (fatty acid-binding proteins) bind saturated and unsaturated long-chain fatty acids reversibly and might be involved in the transport of lipids to specific cellular compartments. FABP5 (fatty acid binding protein 5) serves as a transporter for endocannabinoid. FABP5 is also known as epidermal FABP (E-FABP) or mal1. FABP5 is significantly associated with the development of insulin resistance and atherosclerosis. This gene was found to be overexpressed in cervical cancer and it serves as an important biomarker for cervical cancer. In vitro study proves that FABP5 is necessary for cell proliferation, colony formation, cell migration, and invasion of cervical cancer. FABP5 is associated with cancer types such as bladder, pancreas, prostate, breast cancer and glioblastoma.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fatty acid activated PPAR? promotes tumorigenicity of prostate cancer cells by up regulating VEGF via PPAR responsive elements of the promoter.
Forootan FS
Oncotarget, 7(8), 9322-9339 (2016)
FABP5 correlates with poor prognosis and promotes tumor cell growth and metastasis in cervical cancer.
Wang W
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 37(11), 14873-14883 (2016)
The Antinociceptive Agent SBFI-26 Binds to Anandamide Transporters FABP5 and FABP7 at Two Different Sites.
Hsu HC
Biochemistry, 56(27), 3454-3462 (2017)
Transcriptome and Metabolome Analyses in Exogenous FABP4- and FABP5-Treated Adipose-Derived Stem Cells.
Yamamoto T
PLoS ONE, 11(12), 1-19 (2016)
Takuro Iwao et al.
PloS one, 18(2), e0281946-e0281946 (2023-02-17)
Nutrients are actively taken up by the brain via various transporters at the blood-brain barrier (BBB). A lack of specific nutrients in the aged brain, including decreased levels of docosahexaenoic acid (DHA), is associated with memory and cognitive dysfunction. To

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service