Skip to Content
Merck
All Photos(1)

Key Documents

PZ0141

Sigma-Aldrich

PD-156707

≥98% (HPLC)

Synonym(s):

α-[2-(4-Methoxyphenyl)-2-oxo-1-[(3,4,5-trimethoxyphenyl)methyl]ethylidene]-1,3-benzodioxole-5-acetic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€103.00
25 MG
€372.00

€103.00


Estimated to ship on22 April 2025



Select a Size

Change View
5 MG
€103.00
25 MG
€372.00

About This Item

Empirical Formula (Hill Notation):
C28H25O9·Na
CAS Number:
Molecular Weight:
528.48
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

€103.00


Estimated to ship on22 April 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥20 mg/mL

storage temp.

room temp

SMILES string

COc1ccc(cc1)C(=O)\C(Cc2cc(OC)c(OC)c(OC)c2)=C(/C(=O)O[Na])c3ccc4OCOc4c3

InChI

1S/C28H26O9.Na/c1-32-19-8-5-17(6-9-19)26(29)20(11-16-12-23(33-2)27(35-4)24(13-16)34-3)25(28(30)31)18-7-10-21-22(14-18)37-15-36-21;/h5-10,12-14H,11,15H2,1-4H3,(H,30,31);/q;+1/p-1/b25-20-;

InChI key

ZLHQEGFYBMZQGM-RKVLWQGQSA-M

Biochem/physiol Actions

PD-156707 is a selective endothelin receptor (ETA) antagonist.

Features and Benefits

This compound is featured on the Endothelin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service