Skip to Content
Merck
All Photos(1)

Documents

P9800

Sigma-Aldrich

Octaethylene glycol monotetradecyl ether

Synonym(s):

C14E8, Polyoxyethylene (8) myristyl ether, Tetradecyl octaethylene glycol ether, Tetradecyloctaglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13(OCH2CH2)8OH
CAS Number:
Molecular Weight:
566.81
Beilstein:
5996901
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

non-ionic

mol wt

566.81 g/mol

technique(s)

electrophoresis: suitable
mass spectrometry (MS): suitable
protein quantification: suitable

SMILES string

CCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C30H62O9/c1-2-3-4-5-6-7-8-9-10-11-12-13-15-32-17-19-34-21-23-36-25-27-38-29-30-39-28-26-37-24-22-35-20-18-33-16-14-31/h31H,2-30H2,1H3

InChI key

NHHAZFYVKWSFIR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service