Skip to Content
Merck
All Photos(1)

Key Documents

P0108

Sigma-Aldrich

PRL-3 Inhibitor I

≥98% (HPLC), solid

Synonym(s):

5-[[5-Bromo-2-[(2-bromophenyl)methoxy]phenyl]methylene]-2-thioxo-4-thiazolidinone, Phosphatase of regenerating liver-3, Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11Br2NO2S2
CAS Number:
Molecular Weight:
485.21
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

storage condition

protect from light

color

yellow

solubility

DMSO: >10 mg/mL
H2O: <2 mg/mL

storage temp.

−20°C

SMILES string

Brc1ccc(OCc2ccccc2Br)c(c1)\C=C3\SC(=S)NC3=O

InChI

1S/C17H11Br2NO2S2/c18-12-5-6-14(22-9-10-3-1-2-4-13(10)19)11(7-12)8-15-16(21)20-17(23)24-15/h1-8H,9H2,(H,20,21,23)/b15-8+

InChI key

HXNBAOLVPAWYLT-OVCLIPMQSA-N

Application

PRL-3 Inhibitor I has been used as an inhibitor of phosphatase of regenerating liver-3 (PRL-3):
  • to test its effect on classical Hodgkin lymphoma cell survival
  • in the human umbilical vein endothelial cells tube formation assay
  • to test its effect on the migration of neural crest cells

Biochem/physiol Actions

PRL-3 Inhibitor I is a rhodanine derivative with an IC50 value of 0.9 μM against phosphatase of regenerating liver-3 (PRL-3), a nonclassical protein tyrosine phosphatase that has recently been shown to be involved in cancer metastasis. PRL-3 Inhibitor I reduced the invasiveness of B16F10 melanoma cells in a cell based assay.

Features and Benefits

This compound is featured on the Phosphoprotein Phosphatases (Tyrosine) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianliang Xu et al.
PloS one, 6(11), e27165-e27165 (2011-11-11)
Phosphatase of regenerating liver 3 (PRL-3) is known to be overexpressed in many tumors, and its transcript level is high in the vasculature and endothelial cells of malignant tumor tissue. However, the mechanism(s) underlying its enhanced expression and its function
Magnus Aassved Hjort et al.
Experimental hematology & oncology, 7, 8-8 (2018-04-14)
Phosphatase of regenerating liver-3 (PRL-3) is implicated in oncogenesis of hematological and solid cancers. PRL-3 expression increases metastatic potential, invasiveness and is associated with poor prognosis. With this study, we aimed to show a possible oncogenic role of PRL-3 in
Ying-Qian Lu et al.
Stem cell research, 34, 101354-101354 (2019-01-06)
The human iPS cell line, hiPS-SPG76 (FJMUi001-A), derived from skin fibroblasts from a 42-year-old male hereditary spastic paraplegia patient carrying compound heterozygous p.P498L (c.1493C > T) and p.R618W (c.1852C > T) mutations in the CAPN1 gene, was generated by non-integrative reprogramming vectors encoding OCT3/4
Jin Hee Ahn et al.
Bioorganic & medicinal chemistry letters, 16(11), 2996-2999 (2006-03-15)
A series of rhodanine derivatives was synthesized and evaluated for their ability to inhibit PRL-3. Benzylidene rhodanine derivative showed good biological activity, while compound 5e was the most active in this series exhibiting an IC50 value of 0.9 microM in
E G Garcia et al.
Leukemia, 35(3), 679-690 (2020-07-02)
T-cell acute lymphoblastic leukemia (T-ALL) is an aggressive malignancy of thymocytes and is largely driven by the NOTCH/MYC pathway. Yet, additional oncogenic drivers are required for transformation. Here, we identify protein tyrosine phosphatase type 4 A3 (PRL3) as a collaborating

Articles

Protein tyrosine phosphatases' catalytic mechanism involves transient phosphorylation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service