Skip to Content
Merck
All Photos(1)

Key Documents

N3663

Sigma-Aldrich

Anti-NeuroD1 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-BETA2, Anti-BHF-1, Anti-NDF1, Anti-Neurogenic differentiation factor1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~45 kDa (doublet bands)

species reactivity

human, rat, mouse

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

western blot: 0.5-1 μg/mL using whole extracts of HEK-293T cells expressing human NeuroD1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

NeuroD1 gene is mapped to human chromosome 2q31.3.
Neuronal differentiation 1 (NEUROD1) is a transcription factor which is highly expressed in the brain, pancreas and retina.

Specificity

Anti-NeuroD1 specifically recognizes human, mouse and rat NeuroD1.

Immunogen

synthetic peptide corresponding to amino acids 76-91 of human NeuroD1 conjugated to BSA. The corresponding sequence differs in mouse and rat by two and three amino acids, respectively.

Application

Anti-NeuroD1 antibody produced in rabbit has been used in chromatin immunoprecipitation and immunoblotting.

Biochem/physiol Actions

NeuroD1 is a basic helix-loop-helix (bHLH) transcription factor, which regulates the fate of specific neuronal, endocrine and retinal cells.
Neuronal differentiation 1 (NEUROD1) binds to promoters and regulates gene expression. It can also enhance tumor cell survival in lung tumors. Mutations in the gene encoding NEUROD1 have been associated with neurological problems and early-onset diabetes.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lack of association between NeuroD1/D6 gene polymorphism and heroin dependence in Han-chinese male population
Tsou CC, et al.
Journal of Medical Sciences, 38(5), 228-228 (2018)
Jihan K Osborne et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(16), 6524-6529 (2013-04-05)
Small-cell lung cancer and other aggressive neuroendocrine cancers are often associated with early dissemination and frequent metastases. We demonstrate that neurogenic differentiation 1 (NeuroD1) is a regulatory hub securing cross talk among survival and migratory-inducing signaling pathways in neuroendocrine lung
Winifred P S Wong et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(29), 13057-13062 (2010-07-10)
Estrogen receptors (ERs) protect pancreatic islet survival in mice through rapid extranuclear actions. ERalpha also enhances insulin synthesis in cultured islets. Whether ERalpha stimulates insulin synthesis in vivo and, if so, through which mechanism(s) remain largely unknown. To address these
Feng Wang et al.
Investigative ophthalmology & visual science, 56(1), 150-155 (2014-12-06)
Mutations in the same gene can lead to different clinical phenotypes. In this study, we aim to identify novel genotype-phenotype correlations and novel disease genes by analyzing an unsolved autosomal recessive retinitis pigmentosa (ARRP) Han Chinese family. Whole exome sequencing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service