Skip to Content
Merck
All Photos(1)

Key Documents

M0806

Sigma-Aldrich

Anti-Matrix Metalloproteinase-11, N-Terminal antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MMP-11

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€380.00

€380.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€380.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€380.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1:1,000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MMP11(4320)

Related Categories

Specificity

Reacts with reduced and non-reduced MMP-11. Recognizes active forms of MMP-11. By immunoblotting, the antibody reacts with bands at 58 kDa and 45 kDa.

Immunogen

synthetic peptide corresponding to the N-terminal of active human matrix metalloproteinase-11 (stromelysin-3).

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Physical form

Solution in phosphate buffered saline, pH 7.4 containing 50% glycerol and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denise Rockstroh et al.
Adipocyte, 5(3), 283-297 (2016-09-13)
MicroRNAs (miRNAs) are non-coding RNAs that regulate target gene expression at the post-transcriptional level and are supposed to be implicated in the control of adipogenesis. We aimed to identify miRNAs which are involved in the regulation of human adipogenesis and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service